Zinquin ethyl ester ≥95% (durch HPLC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
2-[[2-Methyl-8-[[(4-methylphenyl)sulphonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester
Z2251-5MG
Z2251-5MGEA
558
EUR
Z2251-5MG
Zinquin ethyl ester ≥95% (durch HPLC), Sigma-Aldrich®
Zinquin ethyl ester
Zinquin ethyl ester. ≥95% (HPLC), solid (2-Methyl-8-[(4-methylphenyl)sulfonylamino]-6-(ethyloxycarbonylmethyloxy)quinoline)
Formel:
C₂₁H₂₂N₂O₅S Lagertemperatur: Kühlschrank |
CAS-Nummer:
181530-09-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...