Microscope, Primostar 3
Lieferant: ZEISS
4155010031000
ZEIS4155010031000EA 3180 EUR
ZEIS4155010031000
Microscope, Primostar 3
Mikroskope
Primostar 3 microscope is a robust and compact microscope for digital teaching and routine lab work.
Transmitted light brightfield, darkfield, simple polarisation, phase contrast and fluorescence contrast are contrast techniques of choice.
Use free of charge labscope app on your Windows PC or iOS device to create images, movies.