Digital camera, Axiocam 305 mono
Lieferant: ZEISS
4265609040000
ZEIS4265609040000EA 6920 EUR
ZEIS4265609040000
Digital camera, Axiocam 305 mono
Kameras
Axiocam 305 mono is a CMOS microscope camera (C-mount) for routine fluorescence applications.
  • 5 MP sensor with 11 mm diagonal for good field of view
  • Fast readout with up to 36 full frame images for good live image experience and easy sample adjustment
  • Dark homogenous background helps to see even the finest structural details
  • Fast USB 3.0 connection, easy to install, with integrated camera power supply
  • Actively temperature stabilised sensor for optimised signal-to-noise and long term stability
  • Fast and efficient operation with ZEN imaging software
Recommended camera adapters: 0,5 × / 0,63 × / 1,0 ×