Face light M8 with light guide
Lieferant: ZEISS
4355080000000
ZEIS4355080000000EA 954 EUR
ZEIS4355080000000
Face light M8 with light guide
Beleuchtung Lichtleiter
Diffuse face light M8 with light guide (D) radiating surface 78×30 mm, outer dimensions 140×58×22 mm, incl. light guide 8/1000 mm and M6/M8 screw adapter.