Wipes for general applications, Multitex® Extra

Lieferant: Zellstoff-Vertriebs-GmbH & Co. KG
11818-00
ZVGA11818-00EA 151 EUR
ZVGA11818-00
Wipes for general applications, Multitex® Extra
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
These high-quality viscose fleece wipes with textile characteristics are suitable for wet and dry cleaning. The cloths are very soft and yet lint-free, they can absorb large amounts of liquid, remain tear-resistant and meet food safety requirements. Multitex® cloths can be washed out after cleaning, wrung out, and reused.

  • Solvent resistant and fibre-free
  • Wipe can take up large amounts of liquid thanks to its coarse hole structure
  • High tear resistance, even when wet

Multitex® cloths can be washed out after cleaning, wrung out, and reused. Ideal for use in sensitive work areas, such as precision mechanics, electrical and automotive industries, and in paint shops.

Colour: White
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR