Tweezer for weights from 50g to 1mg, length 100mm, stainless steel anti-magnetic, tip protected by PVC. 1 * 1 ST
Lieferant: ZWIEBEL
Tweezer for weights from 50g to 1mg, length 100mm, stainless steel anti-magnetic, tip protected by PVC. 1 * 1 ST