Circular cover glasses
Lieferant: VWR International
CSL-103
ZZ49CSL-103EA 0 EUR
ZZ49CSL-103
Circular cover glasses
Deckgläser
These coverslips are designed for hanging-drop, sitting-drop and sandwich-drop crystallisation experiments.
All cover slides can be sealed to the respective crystallisation plates using Bayer silicon grease.