Masterflex® TR-50-TEE-CP Torque Tee Sanitary Clamp Securing Handle, 50 in-lb; 1/EA
Lieferant: Avantor Fluid Handling
Ensure the security of sanitary connections with preset torque output.
- Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
- Easily reach into tight or awkward spaces
- Choose the appropriate torque output per the clamp manufacturer's specifications
- Audible click alerts you when the torque has been reached
- Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
- 'T' shape creates effective leverage for fast, easy one-handed tightening
- Recommended for PTFE, Tuf-Steel®, and Envelope Gaskets
The Rubberfab Torque Tee and universal socket are designed to safely and securely install and remove sanitary clamps. Available with a family of components, the Torque Tee handle creates effective tightening leverage, even in hard-to-reach spaces. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle. The universal socket is required when using the Torque Tee.
<B>Handles require the universal socket (23508-08) to function with standard sanitary clamp wing nuts.</B>