Value Plastics® Fitting, Polypropylene, Straight, Male Luer to Hose Barb Adapter, 1/8" ID; 1000/PK
Lieferant: Avantor Fluid Handling
VWR Item#MFLX30800-24BLK
Your Organization's Price:
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Male luer to ¹/₈" hose barb with 0,778" overall length
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.