Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')europium(III)

Lieferant: Thermo Scientific
Achtung

Synonyme: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III), Eu(TMHD)3

87898.03 87898.06
87898.03EA 73.4 EUR
87898.03 87898.06
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')europium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')europium(III)

CAS No.: 15522-71-1 Molecular Formula: C33H57EuO6 Formula Weight: 701.78 Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 187-189° MDL No.: MFCD00000019

Formel: C₃₃H₅₇EuO₆
Molecular Weight: 701,766 g/mol
Schmelzpunkt: 187…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000019
CAS-Nummer: 15522-71-1
EINECS: 239-565-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR