Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')hafnium(IV) ≥99,7% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)

41909.06 41909.14
41909.06EA 674 EUR
41909.06 41909.14
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')hafnium(IV) ≥99,7% (Metall-Basis)
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')hafnium(IV)

Purity: 99.7% CAS No.: 63370-90-1 Molecular Formula: C44H76HfO8 Formula Weight: 910.5 Physical Form: Powder Melting Point: 315° MDL No.: MFCD01863471

Formel: C₄₄H₇₆HfO₈
Molecular Weight: 910.5 g/mol
Schmelzpunkt: 315 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863471
CAS-Nummer: 63370-90-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR