Bis(2,2,6,6-tetramethyl-2,5-heptandionato-O,O')cobalt(II) ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)
41359.02 41359.04 41359.09
41359.02EA 68.4 EUR
41359.02 41359.04 41359.09
Bis(2,2,6,6-tetramethyl-2,5-heptandionato-O,O')cobalt(II) ≥99,9% (Metall-Basis)
Bis(2,2,6,6-tetramethyl-2,5-heptandionato-O,O')cobalt(II)
Purity: 99.9% CAS No.: 13986-53-3 Molecular Formula: C22H38CoO4 Formula Weight: 425.46 Physical Form: Powder Melting Point: 143° MDL No.: MFCD00233616
Precursor for CVD process
Formel: C₂₂H₃₈CoO₄
Molecular Weight: 425.46 g/mol
Schmelzpunkt: 143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233616
CAS-Nummer: 13986-53-3