Bariumhexafluoracetylacetonat

Lieferant: FLUOROCHEM

Synonyme: Barium hexafluoro-2,4-pentanedionate

004056-1G 004056-5G
FLUO004056-1GEA 48.8 EUR
FLUO004056-1G FLUO004056-5G
Bariumhexafluoracetylacetonat
Bariumhexafluoracetylacetonat
Formel: C₁₀H₂BaF₁₂O₄
Molecular Weight: 551,44 g/mol
MDL: MFCD00075633
CAS-Nummer: 118131-57-0
UN: 1564
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR