Kupfer(II)hexafluoracetylacetonat Hydrat
Lieferant: FLUOROCHEM
Synonyme:
Kupfer(II)Hexafluoro-2,4-pentanedionat Hydrat
009477-1G
009477-5G
FLUO009477-1GEA
79.1
EUR
FLUO009477-1G
FLUO009477-5G
Kupfer(II)hexafluoracetylacetonat Hydrat
Kupfer(II)hexafluoracetylacetonat Hydrat
Formel:
C₁₀H₂CuF₁₂O₄·xH₂O Molecular Weight: 477,64 g/mol (anhy) |
MDL:
MFCD00151019 CAS-Nummer: 155640-85-0 EINECS: 238-850-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...