Hexaamminruthenium(III)chlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride

363340010 363340050 363342500
ACRO363340010EA 161 EUR
ACRO363340010 ACRO363340050 ACRO363342500
Hexaamminruthenium(III)chlorid 98%
Hexaammineruthenium trichloride
Formel: H₁₈Cl₃N₆Ru
Molecular Weight: 309,61 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011478
CAS-Nummer: 14282-91-8
EINECS: 238-176-8
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Elemental Analysis ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR