Hexaamminruthenium(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride
M-Clarity™ Qualitätsstufe = MQ100
262005-1G
262005-250MG
262005-5G
SIAL262005-1GEA
138
EUR
SIAL262005-1G
SIAL262005-250MG
SIAL262005-5G
Hexaamminruthenium(III)chlorid, Sigma-Aldrich®
Hexaammineruthenium trichloride
Formel:
H₁₈Cl₃N₆Ru Molecular Weight: 309,61 g/mol |
MDL:
MFCD00011478 CAS-Nummer: 14282-91-8 EINECS: 238-176-8 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...