Reinecke-Salz Monohydrat ≥93,0% ACS
Lieferant: Thermo Scientific
Achtung
Synonyme: Ammonium reineckate, Ammonium diamminetetrathiocyanatochromate(III) monohydrate
87777.14 87777.22
87777.14EA 79.6 EUR
87777.14 87777.22
Reinecke-Salz Monohydrat ≥93,0% ACS
Reinecke-Salz Monohydrat
Purity: 93.0% min CAS No.: 13573-16-5 Molecular Formula: C4H10CrN7S4.H2O Formula Weight: 354.44 (336.42anhy) Physical Form: Powder Appearance: Dark red Odour: Odourless Solubility: Insoluble in dilute hydrochloric acid. Moderately soluble in water. Soluble in alcohol Merck Reference: 14,8128 UN No.: 3077 MDL No.: MFCD00066638
Used to precipitate primary and secondary amines as their ammonium salts
Formel: C₄H₁₀CrN₇S₄·1H₂O
Molecular Weight: 354,41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066638
CAS-Nummer: 13573-17-6
EINECS: 237-003-3
UN: 3077
ADR: 9,III
Merck Index: 14,8128