Indium-Gallium-Legierung (60:40 gew.%) ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
44240.03
44240.09
44240.18
44240.03EA
38.4
EUR
44240.03
44240.09
44240.18
Indium-Gallium-Legierung (60:40 gew.%) ≥99,99% (Metall-Basis)
Indium-Gallium-Legierung (60:40 gew.%)
Purity: 99.99%
Molecular Formula: Ga:In; 40:60 wt%
Physical Form: Solid
Odour: Odourless
MDL No.: MFCD00144387
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00144387 EINECS: 231-163-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...