Fmoc-D-phenylalanin

Lieferant: BACHEM BIOCHEMICA

BACE4005952.0100EA 963 EUR
BACE4005952.0100 BACE4005952.0005 BACE4005952.0025
Fmoc-D-phenylalanin
Fmoc-D-phenylalanin
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387,4 g/mol
MDL: MFCD00062955
CAS-Nummer: 86123-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR