Fmoc-L-phenylalanin

Lieferant: NOVABIOCHEM

Synonyme: N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine

8.52016.0100 8.52016.0025
04-12-1030-100EA 68.2 EUR
04-12-1030-100 04-12-1030-25
Fmoc-L-phenylalanin
Fmoc-L-phenylalanin
Formel: C₂₄H₂₁NO₄
MDL: MFCD00037128
CAS-Nummer: 35661-40-6
EINECS: 252-661-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR