2-(1,2-Benzisoxazol-3-yl)essigsäure

Lieferant: BIONET RESEARCH

10W-0718.500 10W-0718.1 10W-0718.5 10W-0718.10
BIOT10W-0718.500EA 0 EUR
BIOT10W-0718.500 BIOT10W-0718.1 BIOT10W-0718.5 BIOT10W-0718.10
2-(1,2-Benzisoxazol-3-yl)essigsäure
2-(1,2-Benzisoxazol-3-yl)essigsäure
Formel: C₉H₇NO₃
MDL: MFCD02180388
CAS-Nummer: 4865-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR