2-(2-Thienyl)-1,3-thiazolidin-4-carbonsäure

Lieferant: MENAI (PART OF CHEMPUR)

MENAST3558.5EA 0 EUR
MENAST3558.5 MENAST3558.10 MENAST3558.2 MENAST3558.1
2-(2-Thienyl)-1,3-thiazolidin-4-carbonsäure
2-(2-Thienyl)-1,3-thiazolidin-4-carbonsäure
Formel: C₈H₉NO₂S₂
MDL: MFCD00022489
CAS-Nummer: 32451-19-7
EINECS: 251-055-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR