3-(4-Methylphenyl)-2,1-benzisoxazol-5-carbaldehyd

Lieferant: BIONET RESEARCH

7H-923.500 7H-923.1 7H-923.5 7H-923.10
BIOT7H-923.500EA 0 EUR
BIOT7H-923.500 BIOT7H-923.1 BIOT7H-923.5 BIOT7H-923.10
3-(4-Methylphenyl)-2,1-benzisoxazol-5-carbaldehyd
3-(4-Methylphenyl)-2,1-benzisoxazol-5-carbaldehyd
Formel: C₁₅H₁₁NO₂
MDL: MFCD00663843
CAS-Nummer: 328547-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR