3-Phenyl-2,1-benzisoxazol-5-carbaldehyd

Lieferant: BIONET RESEARCH

7H-911.500 7H-911.1 7H-911.5 7H-911.10
BIOT7H-911.500EA 0 EUR
BIOT7H-911.500 BIOT7H-911.1 BIOT7H-911.5 BIOT7H-911.10
3-Phenyl-2,1-benzisoxazol-5-carbaldehyd
3-Phenyl-2,1-benzisoxazol-5-carbaldehyd
Formel: C₁₄H₉NO₂
MDL: MFCD00623237
CAS-Nummer: 94752-06-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR