5,6-Dimethyl-2,1,3-benzothiadiazol
Lieferant: Thermo Scientific
RDP00075.1
RDP00075.10
MAYBRDP00075.1EA
0
EUR
MAYBRDP00075.1
MAYBRDP00075.10
5,6-Dimethyl-2,1,3-benzothiadiazol
5,6-Dimethyl-2,1,3-benzothiadiazole
Formel:
C₈H₈N₂S Molecular Weight: 164.23 g/mol |
MDL:
MFCD00174277 CAS-Nummer: 1887-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...