2,1,3-Benzothiadiazol-5-ylamin

Lieferant: Thermo Scientific

Synonyme: 5-Amino-2,1,3-benzothiadiazole, Benzo[c][1,2,5]thiadiazol-5-amine

CC09214.250 CC09214DA CC09214.0001
MAYBCC09214.250EA 0 EUR
MAYBCC09214.250 MAYBCC09214DA ABCRCC09214.0001
2,1,3-Benzothiadiazol-5-ylamin
2,1,3-Benzothiadiazol-5-ylamin
Formel: C₆H₅N₃S
Molecular Weight: 151.1900 g/mol
MDL: MFCD00185826
CAS-Nummer: 874-37-3
UN: 2811
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR