4-Chlor-2-(trifluormethyl)chinazolin

Lieferant: Thermo Scientific

CD02952.250 CD02952.1
MAYBCD02952.250EA 0 EUR
MAYBCD02952.250 MAYBCD02952.1
4-Chlor-2-(trifluormethyl)chinazolin
4-Chloro-2-(trifluoromethyl)quinazoline
Formel: C₉H₄ClF₃N₂
Molecular Weight: 232,5900 g/mol
MDL: MFCD00793683
CAS-Nummer: 52353-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR