1-(2-Tetrahydrofuroyl)piperazin

Lieferant: FLUOROCHEM

Synonyme: (Piperazin-1-yl)(tetrahydrofuran-2-yl)methanone

009856-10G 009856-25G
FLUO009856-10GEA 0 EUR
FLUO009856-10G FLUO009856-25G
1-(2-Tetrahydrofuroyl)piperazin
1-(2-Tetrahydrofuroyl)piperazin
Formel: C₉H₁₆N₂O₂
Molecular Weight: 184.24 g/mol
MDL: MFCD00190247
CAS-Nummer: 63074-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR