Isosorbid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D-Isosorbide, (3R,3aR,6S,6aR)-Hexahydrofuro[3,2-b]furan-3,6-diol
M-Clarity™ Qualitätsstufe = MQ100
329207-100G
329207-500G
SIAL329207-100GEA
85.1
EUR
SIAL329207-100G
SIAL329207-500G
Isosorbid, Sigma-Aldrich®
Isosorbid
Formel:
C₆H₁₀O₄ Molecular Weight: 146,14 g/mol |
MDL:
MFCD00064827 CAS-Nummer: 652-67-5 EINECS: 211-492-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...