Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
BOP-Cl
310060050
310060250
ACRO310060050EA
73
EUR
ACRO310060050
ACRO310060250
Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid 97%
Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid
Formel:
C₆H₈ClN₂O₅P Molecular Weight: 254,6 g/mol Schmelzpunkt: 191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010077 CAS-Nummer: 68641-49-6 UN: 1759 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...