Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
BOP-Cl
L08775.06
L08775.14
L08775.06EA
62.9
EUR
L08775.06
L08775.14
Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid ≥97%
Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid
Purity: 97%
CAS No.: 68641-49-6
Molecular Formula: C6H8ClN2O5P
Formula Weight: 254.57
Melting Point: ca 195° dec.
Beilstein Reference: 3654596
UN No.: 3261
MDL No.: MFCD00010077
Formel:
C₆H₈CIN₂O₅P Molecular Weight: 254,6 g/mol Schmelzpunkt: ∼ 195 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010077 CAS-Nummer: 68641-49-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...