Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
BOP-Cl
15140-25G
15140-5G
SIAL15140-25GEA
272
EUR
SIAL15140-25G
SIAL15140-5G
Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid, Sigma-Aldrich®
Bis(2-oxo-3-oxazolidinyl)phosphinsäurechlorid
Formel:
C₆H₈ClN₂O₅P Molecular Weight: 254,6 g/mol |
MDL:
MFCD00010077 CAS-Nummer: 68641-49-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...