6-Piperidinonicotinsäure
Lieferant: BIONET RESEARCH
Synonyme:
6-(Piperidin-1-yl)nicotinic acid, 6-(1-Piperidinyl)pyridine-3-carboxylic acid, 6-(Piperidin-1-yl)pyridine-3-carboxylic acid
6W-0335.500
6W-0335.1
6W-0335.5
6W-0335.10
BIOT6W-0335.500EA
0
EUR
BIOT6W-0335.500
BIOT6W-0335.1
BIOT6W-0335.5
CHMP6W-0335.10
6-Piperidinonicotinsäure
6-Piperidinonicotinsäure
Formel:
C₁₁H₁₄N₂O₂ Molecular Weight: 206.2411 g/mol |
MDL:
MFCD03791231 CAS-Nummer: 120800-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...