6-Piperidinonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-(Piperidin-1-yl)nicotinic acid, 6-(1-Piperidinyl)pyridine-3-carboxylic acid, 6-(Piperidin-1-yl)pyridine-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

702382-1G 702382-250MG
SIAL702382-1GEA 189 EUR
SIAL702382-1G SIAL702382-250MG
6-Piperidinonicotinsäure, Sigma-Aldrich®
6-Piperidinonicotinsäure
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,2411 g/mol
MDL: MFCD03791231
CAS-Nummer: 120800-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR