Pyridino[3,2-b]pyrazin

Lieferant: CHEMPUR

Synonyme: Pyrido[2,3-b]pyrazine

JFD01543.250 JFD01543.1
CHMPJFD01543.250EA 0 EUR
CHMPJFD01543.250 MAYBJFD01543.1
Pyridino[3,2-b]pyrazin
Pyridino[3,2-b]pyrazin
Formel: C₇H₅N₃
Molecular Weight: 131,14 g/mol
MDL: MFCD00006717
CAS-Nummer: 322-46-3
EINECS: 206-294-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR