2-(4-Chlorphenyl)-4-methyl-5-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

12F-341S.500 12F-341S.1 12F-341S.5 12F-341S.10
BIOT12F-341S.500EA 0 EUR
BIOT12F-341S.500 BIOT12F-341S.1 BIOT12F-341S.5 BIOT12F-341S.10
2-(4-Chlorphenyl)-4-methyl-5-thiazolcarbonsäure
2-(4-Chlorophenyl)-4-methyl-5-thiazolecarboxylic acid
Formel: C₁₁H₈ClNO₂S
MDL: MFCD00141960
CAS-Nummer: 54001-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR