4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Lieferant: BIONET RESEARCH
2G-307S.500
2G-307S.1
2G-307S.5
2G-307S.10
BIOT2G-307S.500EA
0
EUR
BIOT2G-307S.500
BIOT2G-307S.1
BIOT2G-307S.5
BIOT2G-307S.10
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Formel:
C₁₀H₈N₂O₂S Molecular Weight: 220,25 g/mol |
MDL:
MFCD01316364 CAS-Nummer: 34418-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...