4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid
1G-380S.500
1G-380S.1
1G-380S.5
1G-380S.10
BIOT1G-380S.500EA
0
EUR
BIOT1G-380S.500
BIOT1G-380S.1
BIOT1G-380S.5
BIOT1G-380S.10
4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure
4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure
Formel:
C₁₀H₈N₂O₂S Molecular Weight: 220.25 g/mol |
MDL:
MFCD00171753 CAS-Nummer: 39091-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...