4-Methyl-2-(2-thienyl)-5-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 4-Methyl-2-(thiophen-2-yl)thiazole-5-carboxylic acid

3G-423S.500 3G-423S.1 3G-423S.5 3G-423S.10
BIOT3G-423S.500EA 0 EUR
BIOT3G-423S.500 BIOT3G-423S.1 BIOT3G-423S.5 BIOT3G-423S.10
4-Methyl-2-(2-thienyl)-5-thiazolcarbonsäure
4-Methyl-2-(2-thienyl)-5-thiazolecarboxylic acid
Formel: C₉H₇NO₂S₂
Molecular Weight: 225.290 g/mol
MDL: MFCD01936004
CAS-Nummer: 209540-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR