[2-(4-Chlorphenyl)-4-methyl-1,3-thiazol-5-yl]methanamin

Lieferant: BIONET RESEARCH

3W-0217.500 3W-0217.1 3W-0217.5 3W-0217.10
BIOT3W-0217.500EA 0 EUR
BIOT3W-0217.500 BIOT3W-0217.1 BIOT3W-0217.5 BIOT3W-0217.10
[2-(4-Chlorphenyl)-4-methyl-1,3-thiazol-5-yl]methanamin
[2-(4-Chlorphenyl)-4-methyl-1,3-thiazol-5-yl]methanamin
Formel: C₁₁H₁₁N₂ClS
MDL: MFCD03791207
CAS-Nummer: 886360-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR