[2-(4-Chlorphenyl)-4-methyl-1,3-thiazol-5-yl]methanol

Lieferant: BIONET RESEARCH

2W-0294.500 2W-0294.1 2W-0294.5 2W-0294.10
BIOT2W-0294.500EA 0 EUR
BIOT2W-0294.500 BIOT2W-0294.1 BIOT2W-0294.5 BIOT2W-0294.10
[2-(4-Chlorphenyl)-4-methyl-1,3-thiazol-5-yl]methanol
[2-(4-Chlorphenyl)-4-methyl-1,3-thiazol-5-yl]methanol
Formel: C₁₁H₁₀NOClS
MDL: MFCD03791195
CAS-Nummer: 317319-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR