2-(4-Pyridinyl)-4-thiazolcarbonsäure

Lieferant: CHEMPUR

SP00001.250 SP00001.1
CHMPSP00001.250EA 0 EUR
CHMPSP00001.250 CHMPSP00001.1
2-(4-Pyridinyl)-4-thiazolcarbonsäure
2-(4-Pyridinyl)-4-thiazolcarbonsäure
Formel: C₉H₆N₂O₂S
Molecular Weight: 206.22 g/mol
MDL: MFCD00171745
CAS-Nummer: 21278-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR