2-(2-Thienyl)-4-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 2-(2-Thienyl)-1,3-thiazole-4-carboxylicacid

3G-388S.500 3G-388S.1 3G-388S.5 3G-388S.10
BIOT3G-388S.500EA 0 EUR
BIOT3G-388S.500 BIOT3G-388S.1 BIOT3G-388S.5 BIOT3G-388S.10
2-(2-Thienyl)-4-thiazolcarbonsäure
2-(2-Thienyl)-4-thiazolecarboxylic acid
Formel: C₈H₅NO₂S₂
Molecular Weight: 211.25 g/mol
MDL: MFCD01936014
CAS-Nummer: 24044-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR