6-Brom-[1,2,4]triazolo[4,3-a]pyridin
Lieferant: COMBI-BLOCKS
Synonyme:
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine
PY-7006-1G
PY-7006-5G
COBBPY-7006-1GEA
76.3
EUR
COBBPY-7006-1G
COBBPY-7006-5G
6-Brom-[1,2,4]triazolo[4,3-a]pyridin
6-Brom-[1,2,4]triazolo[4,3-a]pyridin
Formel:
C₆H₄BrN₃ Molecular Weight: 198,0200 g/mol |
MDL:
MFCD08689506 CAS-Nummer: 108281-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...