6-Brom-8-nitro-[1,2,4]triazolo[4,3-a]pyridin

Lieferant: COMBI-BLOCKS

PY-7181-1G PY-7181-5G
COBBPY-7181-1GEA 375 EUR
COBBPY-7181-1G COBBPY-7181-5G
6-Brom-8-nitro-[1,2,4]triazolo[4,3-a]pyridin
6-Brom-8-nitro-[1,2,4]triazolo[4,3-a]pyridin
Formel: C₆H₃BrN₄O₂
Molecular Weight: 243.200 g/mol
MDL: MFCD09801055
CAS-Nummer: 951884-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR