6-Brom-3-ethyl-3H-[1,2,3]triazolo[4,5-b]pyridin
Lieferant: COMBI-BLOCKS
HI-1471-5G
HI-1471-1G
COBBHI-1471-5GEA
0
EUR
COBBHI-1471-5G
COBBHI-1471-1G
6-Brom-3-ethyl-3H-[1,2,3]triazolo[4,5-b]pyridin
6-Brom-3-ethyl-3H-[1,2,3]triazolo[4,5-b]pyridin
Formel:
C₇H₇BrN₄ |
MDL:
MFCD12913968 CAS-Nummer: 1187386-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...