8-Brom-6-methyl-[1,2,4]triazolo[1,5-a]pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 8-Bromo-6-methyl-[1,2,4]triazolo[1,5-a]pyridine

HI-1294-5G HI-1294-1G
COBBHI-1294-5GEA 690 EUR
COBBHI-1294-5G COBBHI-1294-1G
8-Brom-6-methyl-[1,2,4]triazolo[1,5-a]pyridin
8-Brom-6-methyl-[1,2,4]triazolo[1,5-a]pyridin
Formel: C₇H₆BrN₃
Molecular Weight: 212,0500 g/mol
MDL: MFCD09878402
CAS-Nummer: 957062-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR