5-(4-Boc-piperazino)-2-nitroanisol
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 4-(3-methoxy-4-nitrophenyl)piperazine-1-carboxylate
HC-3090-5G
HC-3090-1G
COBBHC-3090-5GEA
451
EUR
COBBHC-3090-5G
COBBHC-3090-1G
5-(4-Boc-piperazino)-2-nitroanisol
5-(4-Boc-piperazino)-2-nitroanisol
Formel:
C₁₆H₂₃N₃O₅ Molecular Weight: 337,370 g/mol |
MDL:
MFCD09972251 CAS-Nummer: 1017782-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...