1-Amino-4-hydroxy-9,10-anthrachinon 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Amino-4-hydroxyanthraquinone
B24388.06
B24388.14
B24388.22
B24388.06EA
44.7
EUR
B24388.06
B24388.14
B24388.22
1-Amino-4-hydroxy-9,10-anthrachinon 96%
1-Amino-4-hydroxy-9,10-anthrachinon
Purity: 96%
CAS No.: 116-85-8
Molecular Formula: C14H9NO3
Formula Weight: 239.23
Melting Point: 207-209°
MDL No.: MFCD00001223
Formel:
C₁₄H₉NO₃ Molecular Weight: 239,23 g/mol Schmelzpunkt: 207…209 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001223 CAS-Nummer: 116-85-8 EINECS: 204-163-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...