2-(tert-Butyldimethylsilyloxy)-4-bromanisol

Lieferant: COMBI-BLOCKS

Synonyme: (5-Bromo-2-methoxyphenoxy)(tert-butyl)dimethylsilane

OT-0837-1G OT-0837-5G
COBBOT-0837-1GEA 98.3 EUR
COBBOT-0837-1G COBBOT-0837-5G
2-(tert-Butyldimethylsilyloxy)-4-bromanisol
2-(tert-Butyldimethylsilyloxy)-4-bromanisol
Formel: C₁₃H₂₁BrO₂Si
Molecular Weight: 317.2941 g/mol
MDL: MFCD11855959
CAS-Nummer: 177329-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR